KEY466133091 Key Organics / BIONET

4-(5-{[(4-nitrophenyl)methyl]sulfanyl}-1,3,4-oxadiazol-2-yl)pyridine

  • MDL number MFCD02082756
  • Cas Number : 441289-28-7
  • Molecular Weight : 314.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6292

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-136_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart