KEY466133099 Key Organics / BIONET

1-(4-methylphenyl)-2-({5-[4-(trifluoromethyl)phenyl]-1,3,4-oxadiazol-2-yl}sulfanyl)ethan-1-one

  • MDL number MFCD02187286
  • Cas Number : 477857-02-6
  • Molecular Weight : 378.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6307

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-166_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart