KEY466133101 Key Organics / BIONET

6-(4-methylphenyl)-3-[4-(trifluoromethyl)phenyl]-7H-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazine

  • MDL number MFCD02187289
  • Cas Number : 477857-06-0
  • Molecular Weight : 374.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6312

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-176_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart