KEY466133108 Key Organics / BIONET

5-{[4-(3-fluoropropoxy)phenyl]methylidene}-1,3-diazinane-2,4,6-trione

  • MDL number MFCD02187291
  • Cas Number : 477857-23-1
  • Molecular Weight : 292.266

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6332

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-206_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart