KEY466133110 Key Organics / BIONET

tert-butyl 2-{[5-(4-methoxyphenyl)-1,3,4-oxadiazol-2-yl]sulfanyl}acetate

  • MDL number MFCD02082669
  • Cas Number : 477857-27-5
  • Molecular Weight : 322.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6336

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-211_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart