KEY466133113 Key Organics / BIONET

1,2-dimethyl (1R,2R)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate

  • MDL number MFCD08690904
  • Cas Number : 1881275-59-7
  • Molecular Weight : 226.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6339

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-214_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart