KEY466133115 Key Organics / BIONET

2-{[5-(4-tert-butylphenyl)-1,3,4-oxadiazol-2-yl]sulfanyl}-1-(4-methylphenyl)ethan-1-one

  • MDL number MFCD02082769
  • Cas Number : 477857-34-4
  • Molecular Weight : 366.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-222_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart