KEY466133133 Key Organics / BIONET

ethyl 1-phenyl-5-(2-{[(prop-2-en-1-yl)carbamoyl]carbonyl}-1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD02082867
  • Cas Number : 477709-22-1
  • Molecular Weight : 392.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6390

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-351S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart