KEY466133136 Key Organics / BIONET

4-[(E)-[(4-methylphenyl)methylidene]amino]-5-(thiophen-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02102307
  • Cas Number : 474974-63-5
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6418

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-388S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart