KEY466133138 Key Organics / BIONET

4-[(E)-[(3-nitrophenyl)methylidene]amino]-5-(thiophen-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02102310
  • Cas Number : 118159-01-6
  • Molecular Weight : 331.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6423

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-398S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart