KEY466133144 Key Organics / BIONET

methyl 3-{2-[2-oxo-2-(piperidin-1-yl)acetyl]-1H-pyrrol-1-yl}thiophene-2-carboxylate

  • MDL number MFCD02571099
  • Cas Number : 477857-74-2
  • Molecular Weight : 346.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6438

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-516S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart