KEY466133150 Key Organics / BIONET

3-(4-chlorophenyl)-5-(methylsulfanyl)thiophene-2-carboxylic acid

  • MDL number MFCD02083319
  • Cas Number : 203189-46-2
  • Molecular Weight : 284.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6460

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-544S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart