KEY466133152 Key Organics / BIONET

3-{4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD02186982
  • Cas Number : 477858-21-2
  • Molecular Weight : 377.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6505

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-609_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart