KEY466133163 Key Organics / BIONET

bis(4-chlorophenyl)-4H-1,2,4-triazol-4-amine

  • MDL number MFCD02082685
  • Cas Number : 52868-19-6
  • Molecular Weight : 305.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6537

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-656_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart