KEY466133165 Key Organics / BIONET

3-[cyclohexyl(2-hydroxyethyl)amino]-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD02570756
  • Cas Number : 477858-42-7
  • Molecular Weight : 255.281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6542

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-662_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart