KEY466133167 Key Organics / BIONET

2-(1-hydroxy-2-methylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD02082781
  • Cas Number : 107400-33-9
  • Molecular Weight : 205.257

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6546

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-668_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart