KEY466133169 Key Organics / BIONET

3-(4-hydroxyphenyl)-2-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

  • MDL number MFCD02570757
  • Cas Number : 477858-45-0
  • Molecular Weight : 297.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6548

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-670_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart