KEY466133172 Key Organics / BIONET

(2-chlorophenyl)(7-methoxy-1-benzofuran-2-yl)methanol

  • MDL number MFCD02082690
  • Cas Number : 477858-47-2
  • Molecular Weight : 288.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6551

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-677_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart