KEY466133190 Key Organics / BIONET

N'-[(1Z)-1-(4-methylphenyl)ethylidene]-1-(6-methylpyridin-2-yl)-1H-imidazole-4-carbohydrazide

  • MDL number MFCD02082697
  • Cas Number : 477858-70-1
  • Molecular Weight : 333.395

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6592

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-836_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart