KEY466133191 Key Organics / BIONET

methyl 2-{4-hydroxy-2-oxo-1-[(thiophen-2-yl)methyl]-1,2-dihydropyridin-3-yl}acetate

  • MDL number MFCD02187300
  • Cas Number : 477858-72-3
  • Molecular Weight : 279.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6594

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11P-842_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart