KEY466133193 Key Organics / BIONET

N'-(5-iodo-2,6-dimethylpyrimidin-4-yl)-2,2-dimethylpropanehydrazide

  • MDL number MFCD04124338
  • Cas Number : 860649-97-4
  • Molecular Weight : 348.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6599

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart