KEY466133196 Key Organics / BIONET

2-[3-(2,5-dimethyl-1H-pyrrol-1-yl)thiophen-2-yl]-5-methyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD03102468
  • Cas Number : 691868-53-8
  • Molecular Weight : 399.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6606

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart