KEY466133204 Key Organics / BIONET

2-(3,5-dimethyl-1H-pyrazol-1-yl)-4-(3-nitrophenyl)-1,3-thiazole

  • MDL number MFCD02661607
  • Cas Number : 956741-77-8
  • Molecular Weight : 300.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6631

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart