KEY466133208 Key Organics / BIONET

ethyl 6-(4-fluorophenyl)-8-methyl-4-oxo-2H,3H,4H,6H-pyrimido[2,1-b][1,3]thiazine-7-carboxylate

  • MDL number MFCD00695780
  • Cas Number : 254433-14-2
  • Molecular Weight : 348.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6647

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0209_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart