KEY466133210 Key Organics / BIONET

(2-chlorophenyl)methyl N-(3-chloro-1,4-dioxo-1,4-dihydronaphthalen-2-yl)carbamate

  • MDL number MFCD02572004
  • Cas Number : 477858-81-4
  • Molecular Weight : 376.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6653

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0219_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart