KEY466133230 Key Organics / BIONET

ethyl 3-acetyl-6-methyl-4-phenyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD00649136
  • Cas Number : 111535-64-9
  • Molecular Weight : 318.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6703

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0291_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart