KEY466133232 Key Organics / BIONET

1-[3-(4-methoxyphenyl)-2,1-benzoxazol-5-yl]ethan-1-one

  • MDL number MFCD02661618
  • Cas Number : 439094-07-2
  • Molecular Weight : 267.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6715

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0320_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart