KEY466133243 Key Organics / BIONET

2-({1-[4-(trifluoromethoxy)phenyl]-1H-pyrrol-3-yl}methylidene)propanedinitrile

  • MDL number MFCD02661628
  • Cas Number : 439094-26-5
  • Molecular Weight : 303.244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6750

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0636_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart