KEY466133249 Key Organics / BIONET

6-chloro-N-[2-(1H-indol-3-yl)ethyl]pyridazin-3-amine

  • MDL number MFCD02661635
  • Cas Number : 264619-75-2
  • Molecular Weight : 272.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6768

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-0804_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart