KEY466133256 Key Organics / BIONET

(E)-methoxy({[4-(4-methylpiperazin-1-yl)-3-nitrophenyl]methylidene})amine

  • MDL number MFCD04124342
  • Cas Number : 860650-08-4
  • Molecular Weight : 278.312

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Dark Orange Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6792

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-1123_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart