KEY466133257 Key Organics / BIONET

(E)-({4-[4-(4-fluorobenzoyl)piperazin-1-yl]-3-nitrophenyl}methylidene)(methoxy)amine

  • MDL number MFCD03011984
  • Cas Number : 383147-15-7
  • Molecular Weight : 386.383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6795

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-1132_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart