KEY466133258 Key Organics / BIONET

(E)-methoxy({4-[4-(4-methylbenzoyl)piperazin-1-yl]-3-nitrophenyl}methylidene)amine

  • MDL number MFCD03011986
  • Cas Number : 383147-17-9
  • Molecular Weight : 382.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6798

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-1136_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart