KEY466133259 Key Organics / BIONET

(E)-methoxy({3-nitro-4-[4-(thiophene-2-carbonyl)piperazin-1-yl]phenyl}methylidene)amine

  • MDL number MFCD03011988
  • Cas Number : 383147-21-5
  • Molecular Weight : 374.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6800

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-1138_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart