KEY466133261 Key Organics / BIONET

(E)-{[4-(4-methanesulfonylpiperazin-1-yl)-3-nitrophenyl]methylidene}(methoxy)amine

  • MDL number MFCD04124347
  • Cas Number : 860650-10-8
  • Molecular Weight : 342.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6814

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-1152_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart