KEY466133264 Key Organics / BIONET

4-methoxy-N-[4-(1H-pyrrol-1-yl)phenyl]benzene-1-sulfonamide

  • MDL number MFCD03012001
  • Cas Number : 383147-41-9
  • Molecular Weight : 328.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6822

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11R-1184_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart