KEY466133280 Key Organics / BIONET

(Z)-1-[1-(naphthalen-2-yl)ethylidene]-2-phenylhydrazine

  • MDL number MFCD00161070
  • Cas Number : 7150-57-4
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6880

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0231_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart