KEY466133281 Key Organics / BIONET

3-(3,4-dimethoxyphenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02918326
  • Cas Number : 381230-33-7
  • Molecular Weight : 308.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6887

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0241_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart