KEY466133285 Key Organics / BIONET

3-(naphthalen-2-yl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02918303
  • Cas Number : 36640-53-6
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-naphthyl)-1-phenyl-1H-pyrazole-4-carbaldehyde
Inchi Code 1S/C20H14N2O/c23-14-18-13-22(19-8-2-1-3-9-19)21-20(18)17-11-10-15-6-4-5-7-16(15)12-17/h1-14H
Inchi Key DUTZVTXRCLAHHF-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6908

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0263_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart