KEY466133286 Key Organics / BIONET

2-{5-methyl-7-oxo-4H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-6-yl}ethyl 4-cyanobenzoate

  • MDL number MFCD03787400
  • Cas Number : 860650-43-7
  • Molecular Weight : 323.312

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6909

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0265_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart