KEY466133288 Key Organics / BIONET

7-(2H-1,3-benzodioxol-5-yl)-2-(3-methoxyphenyl)-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile

  • MDL number MFCD03617583
  • Cas Number : 685108-55-8
  • Molecular Weight : 370.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6915

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0275_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart