KEY466133290 Key Organics / BIONET

(3Z)-3-(1-{[2-(trifluoromethyl)phenyl]amino}ethylidene)oxolan-2-one

  • MDL number MFCD03617585
  • Cas Number : 685108-57-0
  • Molecular Weight : 271.239

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6921

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0291_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart