KEY466133295 Key Organics / BIONET

ethyl 2-[10-(3,4-dimethoxyphenyl)-7-methyl-1,3,8,12-tetraazatricyclo[7.3.0.0^{2,6}]dodeca-2(6),7,9,11-tetraen-3-yl]acetate

  • MDL number MFCD03787403
  • Cas Number : 860650-48-2
  • Molecular Weight : 396.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6932

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0308_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart