KEY466133301 Key Organics / BIONET

N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]benzenesulfonamide

  • MDL number MFCD02333337
  • Cas Number : 76856-51-4
  • Molecular Weight : 399.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6953

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0346_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart