KEY466133302 Key Organics / BIONET

(2Z)-3-[3-(3,4-dimethoxyphenyl)-1-phenyl-1H-pyrazol-4-yl]prop-2-enoic acid

  • MDL number MFCD03617592
  • Cas Number : 381214-65-9
  • Molecular Weight : 350.374

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6955

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0348_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart