KEY466133308 Key Organics / BIONET

5-[(E)-2-(2-fluorophenyl)diazen-1-yl]-2-sulfanylpyrimidine-4,6-diol

  • MDL number MFCD03305467
  • Cas Number : 1173305-20-8
  • Molecular Weight : 266.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6974

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0622_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart