KEY466133312 Key Organics / BIONET

N'-[(1E)-(5-bromo-2-hydroxyphenyl)methylidene]methoxycarbohydrazide

  • MDL number MFCD01336969
  • Cas Number : 356101-71-8
  • Molecular Weight : 273.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6979

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0627_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart