KEY466133339 Key Organics / BIONET

5,7-dimethyl-3-oxo-2-phenyl-3H-4lambda5-[1,2]diazolo[1,2-a]pyrazol-4-ylium-1-olate

  • MDL number MFCD03305492
  • Cas Number : 76434-58-7
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7021

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0670_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart