KEY466133341 Key Organics / BIONET

5-[(4-chloro-3-nitrophenyl)(hydroxy)methyl]-3-ethyl-1,3-thiazolidine-2,4-dione

  • MDL number MFCD03617596
  • Cas Number : 685108-62-7
  • Molecular Weight : 330.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7025

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0800_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart