KEY466133342 Key Organics / BIONET

1-(1H-indazol-4-yl)-3-methylthiourea

  • MDL number MFCD03617597
  • Cas Number : 685108-63-8
  • Molecular Weight : 206.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7026

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0801_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart