KEY466133343 Key Organics / BIONET

3-benzyl-4-hydroxy-1-[2-(1H-indol-3-yl)ethyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD03848707
  • Cas Number : 1797376-58-9
  • Molecular Weight : 344.414

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7031

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11T-0808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart